2,2'-Dinaphthyl ketone 97%

Supplier: Ambeed
Warning

A112228-100MG A112228-10G A112228-250MG A112228-1G
77148-748EA 37.08 USD
77148-748 77148-750 77148-752 77684-997
2,2'-Dinaphthyl ketone 97%
2,2'-Dinaphthyl ketone

Precaución: for Research Use Only

Formula: C₂₁H₁₄O
MW: 282.34 g/mol
MDL Number: MFCD00183496
CAS Number: 613-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR