S3I-201 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-4-(2-(tosyloxy)acetamido)benzoic acid, NSC 74859

A111782-10MG A111782-1MG A111782-50MG A111782-5MG
77148-736EA 32.96 USD
77148-736 77148-738 77148-740 77148-742
S3I-201 ≥98%
S3I-201

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₇S
MW: 365.36 g/mol
MDL Number: MFCD09907564
CAS Number: 501919-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR