(S)-Methyl 2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Clopidogrel

A111444-1G A111444-250MG A111444-5G
77148-716EA 76.22 USD
77148-716 77148-718 77148-720
(S)-Methyl 2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate 98%
(S)-Methyl 2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetate

Precaución: for Research Use Only

Formula: C₁₆H₁₆ClNO₂S
MW: 321.82 g/mol
MDL Number: MFCD05662337
CAS Number: 113665-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR