2,3-Difluoropyridine-4-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2,3-Difluoropyridin-4-yl)boronic acid

A111167-1G A111167-250MG A111167-5G
77148-702EA 18 USD
77148-702 77148-704 77549-152
2,3-Difluoropyridine-4-boronic acid 98%
2,3-Difluoropyridine-4-boronic acid

Precaución: for Research Use Only

Formula: C₅H₄BF₂NO₂
MW: 158.899 g/mol
MDL Number: MFCD12022339
CAS Number: 1263374-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR