3-(Trifluoromethyl)picolinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethyl)pyridine-2-carboxylic acid, 3-(Trifluoromethy)pyridine-2-carboxylic acid

A110573-10G A110573-250MG A110573-5G A110573-1G A110573-25G
77148-686EA 73.13 USD
77148-686 77148-688 77148-690 77570-590 77683-014
3-(Trifluoromethyl)picolinic acid 97%
3-(Trifluoromethyl)picolinic acid

Precaución: For research use only.

Formula: C₇H₄F₃NO₂
MW: 191.1100 g/mol
MDL Number: MFCD07375386
CAS Number: 87407-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR