GW-441756 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
GW 441756, 3-((1-Methyl-1H-indol-3-yl)methylene)-1,3-dihydro-2H-pyrrolo[3,2-b]pyridin-2-one
A110513-10MG
A110513-1MG
A110513-50MG
A110513-5MG
77148-678EA
30.9
USD
77148-678
77148-680
77148-682
77148-684
GW-441756 98%
GW-441756
Precaución: for Research Use Only
Formula:
C₁₇H₁₃N₃O MW: 275.3 g/mol |
MDL Number:
MFCD17010281 CAS Number: 504433-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...