7-Acetoxyindole 95%

Supplier: Ambeed
Warning

A109794-1G A109794-250MG
77148-658EA 141.11 USD
77148-658 77148-660
7-Acetoxyindole 95%
7-Acetoxyindole

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
MDL Number: MFCD00151953
CAS Number: 5526-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR