5-Bromo-4-chloro-6-methyl-2-(methylthio)pyrimidine 98%

Supplier: Ambeed
Warning

A109476-1G A109476-250MG A109476-5G
77148-646EA 45.32 USD
77148-646 77148-648 77148-650
5-Bromo-4-chloro-6-methyl-2-(methylthio)pyrimidine 98%
5-Bromo-4-chloro-6-methyl-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₆BrClN₂S
MW: 253.54 g/mol
MDL Number: MFCD00834124
CAS Number: 17119-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR