GANT61 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-((2-(Pyridin-4-yl)dihydropyrimidine-1,3(2H,4H)-diyl)bis(methylene))bis(N,N-dimethylaniline)

A109328-10MG A109328-1G A109328-1MG A109328-250MG A109328-50MG A109328-5MG
77148-622EA 28.84 USD
77148-622 77148-624 77148-626 77148-628 77148-630 77148-632
GANT61 98%
GANT61

Precaución: for Research Use Only

Formula: C₂₇H₃₅N₅
MW: 429.6 g/mol
MDL Number: MFCD14635408
CAS Number: 500579-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR