2-(Azetidin-1-yl)ethanamine 98%

Supplier: Ambeed
Danger

Synonyms: 2-(azetidin-1-yl)ethan-1-amine

A109187-1G A109187-250MG A109187-5G
77148-616EA 111.24 USD
77148-616 77148-618 77148-620
2-(Azetidin-1-yl)ethanamine 98%
2-(Azetidin-1-yl)ethanamine

Precaución: for Research Use Only

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
MDL Number: MFCD19205720
CAS Number: 795299-77-3
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR