Vonoprazan fumarate 98%

Supplier: Ambeed
Warning

Synonyms: TAK-438, 1-(5-(2-Fluorophenyl)-1-(pyridin-3-ylsulphonyl)-1H-pyrrol-3-yl)-N-methylmethanamine fumarate

A108580-10MG A108580-1G A108580-250MG A108580-25MG A108580-5MG A108580-100MG A108580-5G
77148-604EA 6.18 USD
77148-604 77148-606 77148-608 77148-610 77148-612 77684-977 77684-978
Vonoprazan fumarate 98%
Vonoprazan fumarate

Precaución: for Research Use Only

Formula: C₂₁H₂₀FN₃O₆S
MW: 461.46 g/mol
MDL Number: MFCD18633280
CAS Number: 881681-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR