Methyl-3-bromo-2,6-difluorobenzoate 98%

Supplier: Ambeed
Warning

A108334-1G A108334-250MG
77148-600EA 47 USD
77148-600 77148-602
Methyl-3-bromo-2,6-difluorobenzoate 98%
Methyl-3-bromo-2,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₂
MDL Number: MFCD09951948
CAS Number: 1378875-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR