2,6-Di-tert-butyl-4-nitrophenol 97%

Supplier: Ambeed
Warning

A108204-10G A108204-1G A108204-250MG A108204-5G
77148-592EA 306.94 USD
77148-592 77148-594 77148-596 77148-598
2,6-Di-tert-butyl-4-nitrophenol 97%
2,6-Di-tert-butyl-4-nitrophenol

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₃
MW: 251.32 g/mol
MDL Number: MFCD00664710
CAS Number: 728-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR