Ethyl 8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylate 98%

Supplier: Ambeed
Warning

A108089-1G A108089-250MG A108089-25G
77148-584EA 63.86 USD
77148-584 77148-586 77570-472
Ethyl 8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylate 98%
Ethyl 8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₄
MW: 228.28 g/mol
MDL Number: MFCD16036567
CAS Number: 24730-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR