1-(6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-2-yl)-N-methyl-1H-pyrazole-4-carboxamide 98%
Supplier: Ambeed
|
Warning
|
A107980-10MG
A107980-1MG
A107980-25MG
A107980-2MG
A107980-50MG
A107980-5MG
77148-572EA
120.51
USD
77148-572
77148-574
77148-576
77148-578
77148-580
77148-582
1-(6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-2-yl)-N-methyl-1H-pyrazole-4-carboxamide 98%
1-(6-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-2-yl)-N-methyl-1H-pyrazole-4-carboxamide
Precaución: for Research Use Only
Formula:
C₁₅H₁₈N₈O₅ MW: 390.35 g/mol |
MDL Number:
MFCD09832544 CAS Number: 313348-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...