SMI-4a 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pim Inhibitor 4a, 5-(3-(Trifluoromethyl)benzylidene)thiazolidine-2,4-dione, (5Z)-5-[[3-(Trifluoromethyl)phenyl]methylene]-2,4-thiazolidinedione, TCS-PIM-1-4a, (Z)-5-(3-Trifluoromethylbenzylidene)thiazolidine-2,4-dione
A107880-10MG
A107880-1G
A107880-1MG
A107880-50MG
A107880-5MG
A107880-100MG
77148-556EA
136.99
USD
77148-556
77148-558
77148-560
77148-562
77148-564
77684-971
SMI-4a 98%
SMI-4a
Precaución: for Research Use Only
Formula:
C₁₁H₆F₃NO₂S MW: 273.23 g/mol |
MDL Number:
MFCD01152003 CAS Number: 327033-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...