L-Hyp-(tBu)-OtBu·HCI 95%

Supplier: Ambeed
Warning

A107461-1G A107461-250MG A107461-5G
77148-540EA 28 USD
77148-540 77148-542 77891-785
L-Hyp-(tBu)-OtBu·HCI 95%
L-Hyp-(tBu)-OtBu·HCI

Precaución: for Research Use Only

Formula: C₁₃H₂₅NO₃·HCl
MDL Number: MFCD01862362
CAS Number: 367453-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR