Torin 2 98%

Supplier: Ambeed
Warning

Synonyms: 9-(6-Aminopyridin-3-yl)-1-(3-(trifluoromethyl)phenyl)benzo[h][1,6]naphthyridin-2(1H)-one

A107433-10MG A107433-1MG A107433-25MG A107433-5MG A107433-50MG
77148-532EA 25 USD
77148-532 77148-534 77148-536 77148-538 77684-970
Torin 2 98%
Torin 2

Precaución: for Research Use Only

Formula: C₂₄H₁₅F₃N₄O
MDL Number: MFCD18782652
CAS Number: 1223001-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR