5-Bromo-6-methyl-2,3-dihydro-1H-inden-1-one 97%

Supplier: Ambeed
Warning

A107244-100MG A107244-250MG A107244-1G
77148-524EA 92.7 USD
77148-524 77148-526 77570-432
5-Bromo-6-methyl-2,3-dihydro-1H-inden-1-one 97%
5-Bromo-6-methyl-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₁₀H₉BrO
MW: 225.08 g/mol
MDL Number: MFCD18647934
CAS Number: 1273655-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR