4,5-Dichloro-2-methoxybenzoic acid 97%

Supplier: Ambeed
Warning

A106803-1G A106803-250MG A106803-5G
77148-518EA 45 USD
77148-518 77148-520 77148-522
4,5-Dichloro-2-methoxybenzoic acid 97%
4,5-Dichloro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O₃
MDL Number: MFCD11045611
CAS Number: 201150-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR