1-NM-PP1 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1NM-PP1(PP1 Analog II), PP1 analog II, 1-(1,1-dimethylethyl)-3-(1-naphthalenylmethyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
A106671-10MG
A106671-1MG
A106671-5MG
77148-512EA
37.08
USD
77148-512
77148-514
77148-516
1-NM-PP1 98%
1-NM-PP1
Precaución: for Research Use Only
Formula:
C₂₀H₂₁N₅ MW: 331.41 g/mol |
MDL Number:
MFCD17010191 CAS Number: 221244-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...