2-Chloro-4-hydrazinylbenzoic acid hydrochloride 95%

Supplier: Ambeed
Warning

A106421-100MG A106421-250MG A106421-1G
77148-502EA 6 USD
77148-502 77148-504 77610-840
2-Chloro-4-hydrazinylbenzoic acid hydrochloride 95%
2-Chloro-4-hydrazinylbenzoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈Cl₂N₂O₂
MDL Number: MFCD25371650
CAS Number: 41112-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR