2-Fluoro-3-nitrobenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 1-(bromomethyl)-2-fluoro-3-nitrobenzene

A106120-100MG A106120-250MG A106120-1G A106120-5G
77148-482EA 28 USD
77148-482 77148-484 77684-962 77901-082
2-Fluoro-3-nitrobenzyl bromide 97%
2-Fluoro-3-nitrobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MW: 234.023 g/mol
MDL Number: MFCD11110170
CAS Number: 946125-65-1
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR