N-Boc-ethylamine 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl N-ethylcarbamate, tert-Butyl ethylcarbamate

A105959-100G A105959-10G A105959-1G A105959-25G A105959-5G
77148-472EA 862.99 USD
77148-472 77148-474 77148-476 77148-478 77148-480
N-Boc-ethylamine 95%
N-Boc-ethylamine

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 145.20 g/mol
MDL Number: MFCD12923020
CAS Number: 38267-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR