4-Tolualdehyde 98%

Supplier: Ambeed
Warning

A105810-1000G A105810-100G A105810-25G A105810-500G
77148-460EA 59 USD
77148-460 77148-462 77148-464 77148-466
4-Tolualdehyde 98%
4-Tolualdehyde

Precaución: for Research Use Only

Formula: C₈H₈O
MDL Number: MFCD00006954
CAS Number: 104-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR