2-Fluoro-3-methylbenzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A105748-100MG A105748-1G A105748-250MG A105748-25G A105748-5G
77148-450EA 5.15 USD
77148-450 77148-452 77148-454 77148-456 77148-458
2-Fluoro-3-methylbenzeneboronic acid pinacol ester 97%
2-Fluoro-3-methylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂
MW: 236.09 g/mol
MDL Number: MFCD18384075
CAS Number: 1192548-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR