Methyl 6-bromo-7-azaindole-3-carboxylate ≥95%

Supplier: Ambeed
Warning

A105658-100MG A105658-1G A105658-250MG A105658-50MG
77148-442EA 285.31 USD
77148-442 77148-444 77148-446 77148-448
Methyl 6-bromo-7-azaindole-3-carboxylate ≥95%
Methyl 6-bromo-7-azaindole-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MW: 255.06 g/mol
MDL Number: MFCD20923471
CAS Number: 1342811-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR