(2,3-Difluorophenyl)acetonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 2,3-Difluorobenzyl cyanide, 2-(2,3-Difluorophenyl)acetonitrile, 2,3-Difluorophenylacetonitrile

A105593-10G A105593-1G A105593-25G A105593-5G
77148-432EA 41.2 USD
77148-432 77148-434 77148-436 77148-438
(2,3-Difluorophenyl)acetonitrile 98%
(2,3-Difluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MW: 153.13 g/mol
MDL Number: MFCD00061277
CAS Number: 145689-34-5
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR