2'-Fluoro-6'-methoxyacetophenone 97%
Supplier: Ambeed
|
Warning
|
A105296-10G
A105296-1G
A105296-250MG
A105296-25G
A105296-5G
77148-424EA
27
USD
77148-424
77148-426
77148-428
77148-430
77900-510
2'-Fluoro-6'-methoxyacetophenone 97%
2'-Fluoro-6'-methoxyacetophenone
Precaución: for Research Use Only
Formula:
C₉H₉FO₂ |
MDL Number:
MFCD00671762 CAS Number: 120484-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...