tert-Butyl ((1R,2S,5S)-2-amino-5-(dimethylcarbamoyl)cyclohexyl)carbamate oxalate 95%

Supplier: Ambeed
Warning

A105279-10G A105279-1G A105279-250MG A105279-25G A105279-5G
77148-414EA 106 USD
77148-414 77148-416 77148-418 77148-420 77148-422
tert-Butyl ((1R,2S,5S)-2-amino-5-(dimethylcarbamoyl)cyclohexyl)carbamate oxalate 95%
tert-Butyl ((1R,2S,5S)-2-amino-5-(dimethylcarbamoyl)cyclohexyl)carbamate oxalate

Precaución: for Research Use Only

Formula: C₁₆H₂₉N₃O₇
MDL Number: MFCD28501698
CAS Number: 1210348-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR