Ethyl phenylphosphinate 95%

Supplier: Ambeed
Warning

A105082-100G A105082-10G A105082-1G A105082-250MG A105082-25G A105082-5G
77148-402EA 191 USD
77148-402 77148-404 77148-406 77148-408 77148-410 77148-412
Ethyl phenylphosphinate 95%
Ethyl phenylphosphinate

Precaución: for Research Use Only

Formula: C₈H₁₁O₂P
MDL Number: MFCD11505905
CAS Number: 2511-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR