2-Deoxy-D-glucose 98%

Supplier: Ambeed
Warning

Synonyms: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal

A104420-1G A104420-1MG A104420-250MG A104420-5G A104420-25G A104420-100G
77148-380EA 10.3 USD
77148-380 77148-382 77148-384 77148-386 77610-810 77684-949
2-Deoxy-D-glucose 98%
2-Deoxy-D-glucose

Precaución: for Research Use Only

Formula: C₆H₁₂O₅
MW: 164.16 g/mol
MDL Number: MFCD00151328
CAS Number: 154-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR