tert-Butyl 2-oxo-8-azaspiro[4.5]decane-8-carboxylate 98%

Supplier: Ambeed
Warning

A104318-250MG A104318-5G
77148-370EA 100 USD
77148-370 77570-260
tert-Butyl 2-oxo-8-azaspiro[4.5]decane-8-carboxylate 98%
tert-Butyl 2-oxo-8-azaspiro[4.5]decane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₃
MDL Number: MFCD18792443
CAS Number: 1250994-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR