Ethyl-3,4-dichlorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3,4-dichlorobenzoate

A104245-100G A104245-10G A104245-1G A104245-25G A104245-5G
77148-360EA 99.91 USD
77148-360 77148-362 77148-364 77148-366 77148-368
Ethyl-3,4-dichlorobenzoate 98%
Ethyl-3,4-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MW: 219.07 g/mol
MDL Number: MFCD06204554
CAS Number: 28394-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR