7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1) 98%

Supplier: Ambeed
Warning

A103852-1G A103852-250MG A103852-5G
77148-334EA 24 USD
77148-334 77148-336 77148-338
7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1) 98%
7-Oxa-2-azaspiro[3.5]nonane oxalate (2:1)

Precaución: for Research Use Only

Formula: C₁₆H₂₈N₂O₆
MDL Number: MFCD22422285
CAS Number: 1523571-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR