5-Nitroquinoline-8-carbonitrile 98%

Supplier: Ambeed
Warning

A102725-1G A102725-5G
77148-318EA 168.92 USD
77148-318 77148-320
5-Nitroquinoline-8-carbonitrile 98%
5-Nitroquinoline-8-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₅N₃O₂
MW: 199.17 g/mol
MDL Number: MFCD27947097
CAS Number: 205046-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR