Cidofovir dihydrate 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(((1-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3-hydroxypropan-2-yl)oxy)methyl)phosphonic acid dihydrate

A102690-100MG A102690-10MG A102690-1MG A102690-50MG A102690-250MG
77148-310EA 71.07 USD
77148-310 77148-312 77148-314 77148-316 77684-936
Cidofovir dihydrate 98%
Cidofovir dihydrate

Precaución: for Research Use Only

Formula: C₈H₁₄N₃O₆P·2H₂O
MW: 315.22 g/mol
MDL Number: MFCD16556167
CAS Number: 149394-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR