Dolutegravir sodium salt ≥99%

Supplier: Ambeed
Warning

A102661-10MG A102661-1MG A102661-25MG A102661-50MG A102661-5MG A102661-1G A102661-250MG A102661-5G A102661-100MG
77148-298EA 17 USD
77148-298 77148-300 77148-302 77148-304 77148-306 77148-308 77548-936 77684-934 77684-935 77891-757
Dolutegravir sodium salt ≥99%
Dolutegravir sodium salt

Precaución: for Research Use Only

Formula: C₂₀H₁₈F₂N₃NaO₅
MDL Number: MFCD28405599
CAS Number: 1051375-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR