MGCD0103 98%

Supplier: Ambeed
Warning

Synonyms: Mocetinostat, N-(2-Aminophenyl)-4-[[[4-(3-pyridinyl)-2-pyrimidinyl]amino]methyl]benzamide, N-(2-Aminophenyl)-4-(((4-(pyridin-3-yl)pyrimidin-2-yl)amino)methyl)benzamide, MG0103

A102453-10MG A102453-1MG A102453-5MG A102453-100MG A102453-1G
77148-290EA 29.87 USD
77148-290 77148-292 77148-294 77684-932 77684-933
MGCD0103 98%
MGCD0103

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₆O
MW: 396.44 g/mol
MDL Number: MFCD10565970
CAS Number: 726169-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR