Dimethyl-4-(4-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate 97%
Supplier: Ambeed
|
Warning
|
A102259-100MG
A102259-1G
A102259-250MG
77148-270EA
16
USD
77148-270
77148-272
77148-274
Dimethyl-4-(4-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate 97%
Dimethyl-4-(4-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₇H₁₈ClNO₄ |
MDL Number:
MFCD00619418 CAS Number: 73257-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...