(1R)-1-[3-(Trifluoromethyl)phenyl]ethylamine 95%

Supplier: Ambeed
Danger

Synonyms: (R)-1-(3-(Trifluoromethyl)phenyl)ethanamine

A101701-100MG A101701-250MG A101701-5G
77148-232EA 53.56 USD
77148-232 77148-234 77148-236
(1R)-1-[3-(Trifluoromethyl)phenyl]ethylamine 95%
(1R)-1-[3-(Trifluoromethyl)phenyl]ethylamine

Precaución: for Research Use Only

Formula: C₉H₁₀F₃N
MW: 189.178 g/mol
MDL Number: MFCD03093005
CAS Number: 127852-30-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR