Farnesyl thiosalicylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Salirasib, 2-(((2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-yl)thio)benzoic acid, FTS

A100859-10MG A100859-1MG A100859-25MG A100859-50MG
77148-190EA 38.11 USD
77148-190 77148-192 77148-194 77148-196
Farnesyl thiosalicylic acid 98%
Farnesyl thiosalicylic acid

Precaución: for Research Use Only

Formula: C₂₂H₃₀O₂S
MW: 358.54 g/mol
MDL Number: MFCD00467723
CAS Number: 162520-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR