VX-765 99%

Supplier: Ambeed
Warning

Synonyms: (S)-1-((S)-2-(4-Amino-3-chlorobenzamido)-3,3-dimethylbutanoyl)-N-((2R,3S)-2-ethoxy-5-oxotetrahydrofuran-3-yl)pyrrolidine-2-carboxamide

A100567-10MG A100567-1MG A100567-5MG
77148-170EA 55.62 USD
77148-170 77148-172 77148-174
VX-765 99%
VX-765

Precaución: for Research Use Only

Formula: C₂₄H₃₃ClN₄O₆
MW: 509 g/mol
MDL Number: MFCD23102811
CAS Number: 273404-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR