Human Serum Albumin (HSA) Powder
Supplier: GeminiBio
800-125P-001 800-125P-005 800-125P-025
76994-316EA 81.18 USD
76994-316 76994-310 76994-314
Human Serum Albumin (HSA) Powder
Sera (Serum)
Human albumin is prepared by cold-alcohol fractionation from pooled human plasma and lyophilized. MW ~66 kDa.
Información de suministro: Ships ambient. Store refrigerated at 2 to 8 °C
Precaución: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.