2-Chloro-4-fluorobenzamide 97%

Supplier: Ambeed
Danger

A998509-1G A998509-250MG A998509-25G A998509-5G
77147-394EA 11.34 USD
77147-394 77147-396 77147-398 77147-400
2-Chloro-4-fluorobenzamide 97%
2-Chloro-4-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅ClFNO
MDL Number: MFCD00042531
CAS Number: 88578-90-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR