6-Bromo-2-methyl-[1,2,4]triazolo[1,5-a]pyrimidine 97%

Supplier: Ambeed
Warning

A996868-100MG A996868-1G A996868-250MG
77147-388EA 45.62 USD
77147-388 77147-390 77147-392
6-Bromo-2-methyl-[1,2,4]triazolo[1,5-a]pyrimidine 97%
6-Bromo-2-methyl-[1,2,4]triazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₅BrN₄
MW: 213.03 g/mol
MDL Number: MFCD16987575
CAS Number: 1250444-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR