4-Chloro-2-ethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A994074-1G A994074-250MG A994074-25G A994074-5G
77147-374EA 15.45 USD
77147-374 77147-376 77147-378 77147-380
4-Chloro-2-ethoxyphenylboronic acid 98%
4-Chloro-2-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₃
MDL Number: MFCD03094940
CAS Number: 850568-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR