MN64 98%

Supplier: Ambeed
Danger

A979266-10MG A979266-1MG A979266-25MG A979266-50MG A979266-5MG
77147-346EA 16.48 USD
77147-346 77147-348 77147-350 77147-352 77147-354
MN64 98%
MN64

Precaución: for Research Use Only

Formula: C₁₈H₁₆O₂
MDL Number: MFCD00829158
CAS Number: 92831-11-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR