2-Chloro-5-(trifluoromethyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-(trifluoromethyl)phenylboronic Acid, Dihydroxy(2-chloro-5-(trifluoromethyl)phenyl)borane, (2-Chloro-5-(trifluoromethyl)phenyl)metaboric acid, 2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 2-Chloro-5-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)

A974476-10G A974476-1G A974476-25G A974476-5G
77147-338EA 57.29 USD
77147-338 77147-340 77147-342 77147-344
2-Chloro-5-(trifluoromethyl)phenylboronic acid 97%
2-Chloro-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
MDL Number: MFCD00797335
CAS Number: 182344-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR