(S)-(-)-Limonene 95%

Supplier: Ambeed
Danger

Synonyms: (-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene

A968589-25G A968589-5G
77147-318EA 21.22 USD
77147-318 77747-726
(S)-(-)-Limonene 95%
(S)-(-)-Limonene

Precaución: for Research Use Only

Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00001558
CAS Number: 5989-54-8
UN: 2319
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR